Learn More
Apexbio Technology LLC Triptolide, 5mg. Cas: 38748-32-2 MFCD: MFCD00210565. IL-2/MMP-3/MMP7/MMP19 inhibitor
SDP
Supplier:  Apexbio Technology LLC A38915
Encompass_Preferred
Triptolide inhibits cytokine-induced gene expression via inhibition of NF-kappaB transcriptional activity. 38748-32-2. MFCD00210565
Catalog No. 50-101-1030