Learn More
Medchemexpress LLC Triptolide-d3 | 38748-32-2 | 91.98% | 363.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY32735S1MG
Encompass_Preferred
Triptolide-d3 is the deuterium labeled Triptolide. Triptolide is a diterpenoid triepoxide extracted from the root of *Tripterium wilfordii* with immunosuppressive, anti-inflammatory, antiproliferative, and antitumour effects. Triptolide is an NF-κB activation inhibitor. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04030