Learn More
Apexbio Technology LLC tris(2,2,2-trifluoroethyl) borate 659-18-7 10mg
SDP
Supplier:  Apexbio Technology LLC B821110
Encompass_Preferred
Tris(2 2 2-trifluoroethyl) borate is an organoboron reagent designed to facilitate direct amide bond formation between carboxylic acids and amines It activates carboxyl groups under thermal or microwave-assisted conditions promoting their condensation into amides without the need for transformation into acid chlorides or esters Based on these chemical properties tris(2 2 2-trifluoroethyl) borate holds research potential in peptide coupling medicinal chemistry drug discovery and other synthetic biomedical applications
Catalog No. 50-000-91496