Learn More
Strem, An Ascensus Company CAS# 14876-47-2. 1g. Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), 99% (99.9%-Fe) [Fe(TMHD)3]. MFCD00067465
SDP
Supplier:  Strem, An Ascensus Company 2639101G
Encompass
CAS# 14876-47-2. 1g. Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), 99% (99.9%-Fe) [Fe(TMHD)3]. MFCD00067465. Molecular Weight: 605.66. Molecular Formula: Fe(C11H19O2)3. Color/form: red xtl. Strem# 26-3910. http://www.strem.com/catalog/v/26-3910/
Catalog No. 50-901-11135