Learn More
Strem, An Ascensus Company CAS# 91608-15-0. 10g. Tris(2,4,6-trimethoxyphenyl)phosphine, min. 97%. MFCD00145467
SDP
Supplier:  Strem, An Ascensus Company 15801510G
Encompass
CAS# 91608-15-0. 10g. Tris(2,4,6-trimethoxyphenyl)phosphine, min. 97%. MFCD00145467. Molecular Weight: 532.54. Molecular Formula: [(CH3O)3C6H2]3P. Color/form: light yellow pwdr. Strem# 15-8015. http://www.strem.com/catalog/v/15-8015/
Catalog No. 50-901-10534