Learn More
Strem, An Ascensus Company CAS# 31570-04-4. 250g. Tris(2,4-di-t-butylphenyl)phosphite, 98%. MFCD00072706
SDP
Supplier:  Strem, An Ascensus Company 157720250G
Encompass
CAS# 31570-04-4. 250g. Tris(2,4-di-t-butylphenyl)phosphite, 98%. MFCD00072706. Molecular Weight: 646.93. Molecular Formula: [[(CH3)3C]2C6H3O]3P. Color/form: white pwdr. Strem# 15-7720. http://www.strem.com/catalog/v/15-7720/
Catalog No. 50-901-10497