Learn More
Medchemexpress LLC Tris(2,4-di-tert-butylphenyl) phosphate | 95906-11-9 | MFCD29918884 | 99.7% | 662.92 g/mol | C42H63O4P | 5 MG

Supplier: Medchemexpress LLC HY136177R5MG
Tris(2,4-di-tert-butylphenyl) phosphate is an organophosphorus compound supplied as a high-purity analytical standard for research and analytical applications. It is commonly used as a polymer processing stabilizer and reference material (CAS 95906-11-9).
- High purity (99.7%).
- Molecular weight 662.92 g/mol.
- Chemical formula C42H63O4P.
- White to off-white solid form.
- Commonly used as an analytical standard and reference material.
- Suitable for polymer additive research and enzymatic assay studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.