Learn More
Medchemexpress LLC Tris(2-(4-fluorophenyl)pyridine)iridium | 370878-69-6 | MFCD70878696 | 98.0% | 708.75 g/mol | C33H21F3IrN3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0340415MG
Encompass_Preferred
Ir(p-F-ppy)3 is an iridium(III) cyclometalated complex used as a photocatalyst in photoredox chemistry and as a phosphorescent emitter in optoelectronic research. It facilitates electron-transfer processes in organic synthesis and is applied in studies aimed at improving display and photovoltaic device performance.
  • High photocatalytic activity for photoredox reactions.
  • Suitable as a phosphorescent emitter in optoelectronic studies.
  • Supplied at 98.0% purity for reproducible results.
  • Available in milligram-scale quantities for research applications.
  • Chemical formula C33H21F3IrN3; molecular weight 708.75 g/mol.
Catalog No. 50-002-99468