Learn More
Medchemexpress LLC Tris(2-chloroethyl) phosphate | 115-96-8 | MFCD00000967 | 97.1% | 285.49 g·mol⁻¹ | C6H12Cl3O4P | 10 MG

Supplier: Medchemexpress LLC HY10785910MG
Tris(2-chloroethyl) phosphate (TCEP) is an organophosphate triester provided as a research-grade reagent for laboratory use. It is commonly used as a flame retardant, plasticizer, and chemical intermediate. The material is supplied as a colorless to light-yellow liquid with reported purity ~97.1% and molecular weight 285.49 g·mol⁻¹. For research use only; not for human or clinical use.
- Research-grade purity: 97.1%.
- Physical form: colorless to light-yellow liquid.
- Molecular weight: 285.49 g·mol⁻¹.
- Common applications: flame retardant, plasticizer, chemical intermediate.
- Storage guidance: stable at room temperature; extended stability in solvent at -20 °C to -80 °C.
- Available in small pack sizes, including 10 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.