Learn More
Medchemexpress LLC Tris(2-chloroethyl)phosphate-d12 | 1276500-47-0 | 297.56 | C6D12Cl3O4P | 1 MG

Supplier: Medchemexpress LLC HY107859S1MG
Tris(2-chloroethyl)phosphate-d12 is the deuterium-labeled analogue of tris(2-chloroethyl) phosphate, supplied as an isotope-labeled reference standard for analytical and research applications, particularly as an internal standard for mass spectrometry. It is provided as a purified solid for laboratory use and is intended for research use only.
- Deuterium-labeled internal standard for quantitative analysis.
- Molecular formula C6D12Cl3O4P; molecular weight 297.56.
- High isotopic enrichment suitable for mass spectrometry.
- Storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
- Typical packaging: 1 mg vial for trace-level applications.
- For research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.