Learn More
Medchemexpress LLC Tris(2-chloroethyl)phosphate-d12 | 1276500-47-0 | MFCD20257118 | 99.2% | 297.56 g/mol | C6D12Cl3O4P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY107859S5MG
Encompass_Preferred
Tris(2-chloroethyl)phosphate-d12 is a deuterium-labeled analog of tris(2-chloroethyl) phosphate intended for use as a tracer and internal standard in analytical chemistry. It is provided as a high-purity, isotopically labeled reference material (C6D12Cl3O4P, MW 297.56) for quantitative method development and validation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • High isotopic and chemical purity for reliable quantitation.
  • Colorless to light yellow liquid physical form for easy handling.
  • Stable when stored under recommended temperature conditions.
Catalog No. 50-002-40826