Learn More
Medchemexpress LLC Tris(2-phenylpyridinato-C2,N)iridium(III) | 94928-86-6 | 654.78 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW007596100MG
Encompass_Preferred
Tris(2-phenylpyridinato-C2,N)iridium(III) (Ir(ppy)3) is an iridium(III) containing organometallic complex used as a phosphorescent light-emitting material. It demonstrates high luminous efficiency and good thermal stability, making it suitable for organic optoelectronics, particularly for improving the luminescence efficiency and stability of organic light-emitting diodes (OLEDs).
  • Iridium(III)-containing organometallic complex
  • Used as a phosphorescent light-emitting material
  • Demonstrates high luminous efficiency
  • Exhibits good thermal stability
  • Suitable for organic optoelectronics
  • Enhances luminescence efficiency of OLEDs
  • Improves stability of OLEDs
Catalog No. 50-004-46382