Learn More
Medchemexpress LLC Tris(2-phenylpyridinato-C2,N)iridium(III) | 94928-86-6 | 654.78 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW007596
Encompass_Preferred
Tris(2-phenylpyridinato-C2,N)iridium(III) (Ir(ppy)3) is an iridium(III) containing organometallic complex that functions as a phosphorescent light-emitting material. It exhibits high luminous efficiency and good thermal stability, making it suitable for use in organic optoelectronics, particularly for enhancing the luminescence efficiency and stability of organic light-emitting diodes (OLEDs).
  • Organometallic complex
  • Phosphorescent light-emitting material
  • High luminous efficiency
  • Good thermal stability
  • Suitable for organic optoelectronics
  • Improves luminescence efficiency of OLEDs
  • Improves stability of OLEDs
Catalog No. 50-004-46716