Learn More
Medchemexpress LLC Tris(2-phenylpyridinato-C2,N)iridium(III) | 94928-86-6 | MFCD12022527 | 654.78 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW00759650MG
Encompass_Preferred
Tris(2-phenylpyridinato-C2,N)iridium(III) (Ir(ppy)3) is an organometallic complex containing iridium(III) that serves as a phosphorescent light-emitting material. It offers high luminous efficiency and good thermal stability, making it suitable for use in organic optoelectronics, particularly for enhancing the luminescence efficiency and stability of organic light-emitting diodes (OLEDs).
  • Phosphorescent light-emitting material
  • High luminous efficiency
  • Good thermal stability
  • Suitable for organic optoelectronics
  • Enhances luminescence efficiency and stability of OLEDs
Catalog No. 50-004-45971