Learn More
Medchemexpress LLC Tris(2-phenylpyridinato-C2,N)iridium(III) (Ir(ppy)3) | 94928-86-6 | 654.78 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW007596500MG
Encompass_Preferred
Tris(2-phenylpyridinato-C2,N)iridium(III) (Ir(ppy)3) is an iridium(III)-containing organometallic complex used as a phosphorescent light-emitting material. This compound exhibits high luminous efficiency and good thermal stability, making it suitable for organic optoelectronics, particularly for improving the luminescence efficiency and stability of organic light-emitting diodes (OLEDs).
  • High luminous efficiency
  • Good thermal stability
  • Can be used in organic optoelectronics
  • Especially effective in improving the luminescence efficiency and stability of organic light-emitting diodes (OLEDs)
Catalog No. 50-004-46482