Learn More
Medchemexpress LLC Tris(3-hydroxypropyltriazolylmethyl)amine | 760952-88-3 | MFCD27665386 | 99.0% | 434.50 | C18H30N10O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0210421G
Encompass_Preferred
Tris(3-hydroxypropyltriazolylmethyl)amine (THPTA) is a tris-triazole ligand that stabilizes Cu(I) in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions. It enhances catalytic efficiency and minimizes oxidative side reactions, making it suitable for bioconjugation, oligonucleotide labeling, and other bioorthogonal chemistry applications.
  • Stabilizes Cu(I) for efficient copper-catalyzed azide-alkyne cycloaddition.
  • Improves reaction rates and selectivity in click chemistry.
  • Reduces oxidative degradation and unwanted side reactions.
  • Compatible with bioconjugation and oligonucleotide labeling workflows.
  • Supplied as a solid in laboratory-scale pack sizes for bench use.
Catalog No. 50-003-00888