Learn More
Medchemexpress LLC Tris(3-hydroxypropyltriazolylmethyl)amine | 760952-88-3 | MFCD27665386 | 99.0% | 434.50 g/mol | C18H30N10O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW021042500MG
Encompass_Preferred
Tris(3-hydroxypropyltriazolylmethyl)amine (THPTA) is a triazole-based ligand used to stabilize and accelerate copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) reactions. It improves reaction rates and selectivity for bioconjugation and synthetic applications and is supplied as a high-purity solid for laboratory use.
  • Stabilizes copper(I) to enable efficient CuAAC reactions.
  • Accelerates reaction rates and improves selectivity.
  • High purity suitable for research applications.
  • Available in multiple small-scale package sizes for lab use.
  • White to light brown solid, easy to handle and store.
Catalog No. 50-003-01327