Learn More
Medchemexpress LLC Tris[3-(trimethoxysilyl)propyl] isocyanurate | 26115-70-8 | MFCD00054746 | >95.0% | 615.85 | C21H45N3O12Si3 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW14088325G
Encompass_Preferred
Tris[3-(trimethoxysilyl)propyl] isocyanurate is a trialkoxysilyl-functional isocyanurate used as a coupling agent and adhesion promoter in coatings, adhesives, and polymer formulations. It supplies silane reactivity for bonding between organic and inorganic phases and can serve as a crosslinker to improve mechanical and thermal performance in treated materials. It is provided for research and industrial formulation use.
  • Provides three trimethoxysilyl functional groups for silane coupling.
  • Promotes adhesion between organic polymers and inorganic substrates.
  • Acts as a crosslinking agent to enhance mechanical strength.
  • Improves thermal and chemical resistance of treated materials.
  • Stable liquid suitable for formulation and surface treatment.
  • Recommended low-temperature storage to maintain stability.
Catalog No. 50-002-83484