Learn More
Medchemexpress LLC Tris[3-(trimethoxysilyl)propyl] isocyanurate | 26115-70-8 | MFCD00054746 | 95.0% | 615.85 g/mol | C21H45N3O12Si3 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW14088350G
Encompass_Preferred
Tris[3-(trimethoxysilyl)propyl] isocyanurate is a trifunctional organosilane reagent used as a silane coupling agent and crosslinker for polymer and surface modification applications. The molecule has three 3-(trimethoxysilyl)propyl arms attached to an isocyanurate core, providing hydrolyzable alkoxysilane groups that condense to form siloxane networks. CAS 26115-70-8; molecular weight 615.85 g/mol; physical form: solid-liquid mixture.
  • Provides three reactive alkoxysilane groups for network formation.
  • Improves adhesion between organic polymers and inorganic substrates.
  • Functions as a crosslinker to increase network density and thermal stability.
  • Enables surface functionalization and silanization of substrates.
  • Available in common laboratory pack sizes for formulation and research use.
Catalog No. 50-002-83584