Learn More
Cayman Chemical TrIs 4 7dIphenyl1 10phen 1mg
SDP
Supplier:  Cayman Chemical 161171 MG
Encompass_Preferred
A luminescent probe (absorption λmax: 455 nm, luminescence λmax: 613 nm) widely used for detection and quantitation of oxygen; strongly reduced by molecular oxygen as a result of dynamic quenching; thus oxygen detection can be based on either measurement of intensity or decay time
Catalog No. 50-297-3183