Learn More
Medchemexpress LLC Tris(4-methoxyphenyl)phosphine | 855-38-9 | 99.3% | 352.36 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01377210G
Encompass_Preferred
Tris(4-methoxyphenyl)phosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is employed in Suzuki reaction and also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
  • Biochemical reagent
  • Biological material or organic compound for life science research
  • Employed in Suzuki reaction
  • Used in kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin
Catalog No. 50-004-54541