Learn More
Research Products International Corp TRIS Amino Ultra PC 10 KG
SDP
Supplier:  Research Products International Corp T6004410000.0
Encompass_Preferred
TRIS Amino Ultra PC is a specialized reagent utilized particularly in molecular biology and biochemistry. It is a high-quality, ultra-pure form of TRIS (tris(hydroxymethyl)aminomethane), a commonly used buffering agent in biological applications. 
TRIS Amino Ultra PC is meticulously purified to ensure the absence of contaminants that could interfere with experimental results. Its exceptional purity makes it ideal for use in sensitive assays, such as nucleic acid purification, protein biochemistry, and molecular biology techniques like electrophoresis and Western blotting. 
By providing a stable pH environment, TRIS Amino Ultra PC helps to maintain the integrity and functionality of biological molecules, enabling accurate and reproducible experimental outcomes.
Catalog No. 50-223-0478