Learn More
Research Products International Corp TRIS BASE, 40% SOLUTION, 4 LITER
SDP
Supplier:  Research Products International Corp T600434000.0
Encompass_Preferred
A 40% weight by volume solution of TRIS (3.3M TRIS Base) prepared in Molecular Grade Water.
Catalog No. 50-136-8665