Learn More
Alkali Scientific Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 5 Kg
SDP
Supplier:  Alkali Scientific ISB007CP
Encompass_Preferred
Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 5kg, is a high-purity reagent used in a variety of biochemical and molecular applications, particularly for preparing buffer solutions. Tris Base is essential for maintaining a stable pH in biological and chemical experiments, which makes it indispensable in protein analysis, electrophoresis, and enzyme activity studies. The 5kg size is ideal for larger laboratories or those involved in frequent buffer preparation. Its ultra-pure quality ensures minimal interference with sensitive experiments, making it suitable for protein purification, DNA analysis, and other molecular biology techniques where precise pH control is necessary.
Catalog No. 50-006-59707