Learn More
Alkali Scientific Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 500 G
SDP
Supplier:  Alkali Scientific ISB006CP
Encompass_Preferred
Tris Base Ultra Pure [Tris (Hydroxymethyl) Aminomethane], 500g, is a high-quality reagent used extensively in molecular biology and biochemistry laboratories. This ultra-pure formulation ensures minimal contaminants, making it ideal for experiments that require precise pH control and minimal interference from impurities. Tris Base is commonly used to prepare Tris-buffered solutions for protein purification, enzyme assays, and nucleic acid research. The 500g size is perfect for mid-sized laboratories or for researchers requiring consistent buffer preparation for electrophoresis, PCR, and other biochemical assays. Its ability to maintain pH stability in a wide range of conditions makes it a versatile reagent in the lab.
Catalog No. 50-006-59706