Learn More
Bioworld Tris Buffered Saline Packs, 40 PK(s)
Supplier:  Bioworld 402200882
Encompass_Preferred
Tris Bufferd Saline Packs An ideal all-purpose binding buffer.Each pack yields 500 ml of 25 mM Tris, 0.15 M NaCl, pH 7.2 when dissolved in 500 ml deionized water (total 5 liters). Quantity: 10 packs
Catalog No. 50-255-480