Learn More
Sigma Aldrich Fine Chemicals Biosciences Tris buffered saline with Tween(R) 20 BioUltra, tablet (for 500 mL), pH 7.6 | 10TAB
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 9141410TAB
Encompass
Tris buffered saline with Tween(R) 20 BioUltra, tablet (for 500 mL), pH 7.6 | 10TAB
Catalog No. NC2125001