Learn More
Kd Medical Inc TRIS-GLYCINE BUFFER 10X
SDP
Supplier:  Kd Medical Inc RGF3391
Encompass_Preferred
10X Tris Glycine is used for electrophoretic transfer of protein from polyacrylamide gels to nitrocellulose sheets. 10X concentrated Tris Glycine is ready to dilute to a 1x working solution. One 1liter bottle.
Catalog No. 50-101-8897