Learn More
Alkali Scientific Tris Glycine Buffer 10x Powder, 10 L
SDP
Supplier:  Alkali Scientific TGB002CP
Encompass
Tris Glycine Buffer 10X Powder is a highly efficient buffer concentrate designed for use in protein electrophoresis and other applications requiring stable pH conditions. The 10-liter solution, made from 1702.9 grams of powder, provides an economical option for laboratories with high-volume electrophoresis needs. This buffer ensures consistent protein migration by maintaining a stable pH throughout the electrophoresis process, which is crucial for obtaining reproducible results. It is commonly used in Western blotting, SDS-PAGE, and other gel electrophoresis methods, helping to achieve high-quality separation of proteins and nucleic acids in molecular biology and biochemistry experiments.
Catalog No. 50-006-60923