Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)AMINOMETHAN
SDP
Supplier:  Crescent Chemical Co Inc SE37190.03
Encompass_Preferred
Tris(hydroxymethyl)aminomethane research grade USP. Suitable for preparative purposes and for chromatography. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 37190.03 CAS 77-86-1
Catalog No. 50-248-235