Learn More
Crescent Chemical Co Inc TRIS(HYDROXYMETHYL)AMINOMETHAN
SDP
Supplier:  Crescent Chemical Co Inc SE37180.05
Encompass_Preferred
Tris(hydroxymethyl)aminomethane analytical grade, USP Buffer quality for biological and enzymatic research. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 37180.05 CAS 77-86-1
Catalog No. 50-248-228