Learn More
Medchemexpress LLC Tris (O-TBDMS)3 | 102522-47-4 | MFCD22574795 | >98.0% | 463.92 g·mol^-1 | C22H53NO3Si3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW190935250MG
Encompass_Preferred
Tris (O-TBDMS)3 is a tris-based chemical linker in which three hydroxyl groups are protected as tert-butyldimethylsilyl (TBDMS) ethers. The TBDMS groups are acid-labile, enabling controlled removal under mild acidic conditions. This reagent is used as a protected tris-functional building block in organic synthesis and for preparing conjugation-ready intermediates.
  • Three hydroxyl groups protected as TBDMS ethers.
  • Acid-labile protecting groups allow mild deprotection conditions.
  • Useful as a tris-functional linker for multi-step synthesis.
  • High purity suitable for synthetic applications.
Catalog No. 50-002-81579