Learn More
Medchemexpress LLC Tris(perfluorophenyl)borane | 1109-15-5 | 98.1% | 511.98 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW008525
Encompass_Preferred
Tris(perfluorophenyl)borane is a biochemical reagent used as a biomaterial or organic compound for life science research. It exhibits Lewis acidity comparable to BF3 and outperforms BCl3 and BF3 in both stability and functional group tolerance.
  • Catalyzes reduction of alcohols, carbonyls, or carbonyl-like derivatives alongside a silane reductant
  • Catalyzes boration, hydrogenation, polymerization, hydrosilylation, alcohol or carbonyl deoxygenation, and frustrated Lewis pair chemistry
Catalog No. 50-003-98971