Learn More
Strem, An Ascensus Company CAS# 80510-03-8. 1g. Tris(pyrazol-1-yl)methane, min. 98%. MFCD00040255
SDP
Supplier:  Strem, An Ascensus Company 0732001G
Encompass
CAS# 80510-03-8. 1g. Tris(pyrazol-1-yl)methane, min. 98%. MFCD00040255. Molecular Weight: 214.23. Molecular Formula: C10H10N6. Color/form: white pwdr. Strem# 07-3200. http://www.strem.com/catalog/v/07-3200/
Catalog No. 50-901-09149