Learn More
Alkali Scientific TRIS Tricine SDS Running Buffer 10X Solution, 1 L
Supplier: Alkali Scientific ESD001CP
TRIS Tricine SDS Running Buffer 10X Solution is a concentrated buffer solution used in electrophoresis for running SDS-PAGE gels. This 1L bottle provides enough buffer for multiple runs, and the 10X concentration ensures convenient dilution to the appropriate working concentration. The TRIS-Tricine formulation is known for enhancing the resolution of proteins during electrophoresis, particularly for small proteins. This buffer also stabilizes proteins, ensuring reliable and consistent separation. It is commonly used in proteomics, molecular biology, and biochemistry for high-quality protein analysis and Western blotting applications.