Learn More
Medchemexpress LLC Trisbenzyltriazolylm 500mg | 510758-28-8 | 530.6 g/mol | C30H30N10 | 500 MG

Supplier: Medchemexpress LLC HY116677500MG
Tris(benzyltriazolylmethyl)amine (TBTA) is a tertiary amine ligand containing three 1,2,3-triazole groups. It complexes with and stabilizes copper(I) to accelerate azide-alkyne cycloadditions (click chemistry), and is supplied as a high-purity crystalline solid for use as a biochemical tool in protein and enzyme tagging.
- Complexes with and stabilizes copper(I) to support Cu(I)-catalyzed reactions.
- Accelerates azide-alkyne cycloadditions (click chemistry).
- Supplied as a crystalline solid.
- High purity (≥98%).
- Molecular formula C30H30N10; molecular weight 530.6 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.