Learn More
Medchemexpress LLC Tristearin-d105 | 125941-88-0 | 99.1% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY127035S1MG
Encompass_Preferred
Tristearin-d105 is a deuterium-labeled Tristearin, which is a triglyceride derived from three units of stearic acid. Intended for research use only, it is not sold for patient use. This compound can be used as a tracer or an internal standard in quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-38673