Learn More
Medchemexpress LLC TRITC-conjugated AffiniPure Goat Anti-Mouse IgG H&L | 100 UL

Supplier: Medchemexpress LLC HYP81008100UL
TRITC-conjugated Goat Anti-Mouse IgG H&L is a TRITC-conjugated, goat-derived anti-mouse IgG antibody that conjugates the light and heavy chains of mouse IgG antibodies. It is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, with a pH of 7.3.
- Goat host
- Mouse reactivity
- TRITC conjugation
- Supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide
- Stable for 1 year when stored at -20°C
- Protect from light and avoid repeated freeze/thaw cycles
- Liquid appearance
- Recommended for ICC/IF applications at 1:100-1:1000 dilution
- Recommended for FC applications at 1:500-1:1000 dilution
- Ships with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.