Learn More
Alkali Scientific Triton Lysis Buffer Solution, 500 Milliliters
SDP
Supplier:  Alkali Scientific RIT002CP
Encompass_Preferred
Triton Lysis Buffer Solution, 500 milliliters, is designed for efficient cell lysis and protein extraction in various laboratory applications. This buffer uses Triton X-100, a non-ionic detergent, to solubilize cell membranes without denaturing proteins, making it ideal for obtaining high-quality protein samples for further analysis. The solution is highly effective for disrupting cell structures, allowing the release of proteins, nucleic acids, and other cellular components. It is widely used in molecular biology, immunoprecipitation, and protein extraction workflows. The 500mL volume is convenient for small to medium-scale experiments, ensuring reproducible results and reliable protein extraction for use in enzyme assays, Western blotting, and other biochemical techniques.
Catalog No. 50-006-61705