Learn More
MSC Triton Products Pegboard Storage Board, Steel, 42.5 x 24 in, Blue
Supplier:  MSC 69933737
Encompass
This pegboard storage board is constructed from steel and features a 2-panel system with blue, square holes. Designed for wall mounting, it has a formed panel construction and includes mounting hardware. The epoxy finish ensures durability, and it offers a load capacity of 400 lbs.
  • Mounts to wall
  • Features a two-panel system
  • Constructed from durable steel
  • Has an epoxy finish
  • Includes all necessary mounting hardware
  • Provides 14.0 sq ft of surface area
  • Supports a load capacity of 400 lbs
Catalog No. 50-005-30820