Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cayman Chemical Triton™ X-100 Stock Solution (20% v/v), 250 ml
SDP

Supplier:  Cayman Chemical 600217250/DUP

Encompass_Preferred

Triton™ X-100 Stock Solution (20% v/v), 250 ml

Catalog No. NC2020117

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.