Learn More
Sigma Aldrich Fine Chemicals Biosciences TritonTM X-114 laboratory
Supplier:  Sigma Aldrich Fine Chemicals Biosciences X1141GA
Encompass
Triton(TM) X-114 is a nonionic detergent. It is a good detergent for solubilizing membranes having reversible phase-separation property useful for the separation of intrinsic membrane proteins. Solution of Triton(TM) X-114 is homogeneous at 0C but phase separation into aqueous phase and a detergent phase occurs above 20C. Phase separation of proteins in solutions of Triton(TM) X-114 has been investigated. This octylphenol ethoxylate is compatible with anionic, cationic, and other nonionic surfactants and chemically stable in most acidic alkaline solutions.
Catalog No. 50-289-1457