Learn More
Medchemexpress LLC Tritriacontane (N-tritriacontane) | 630-05-7 | MFCD00010187 | 98.0% | 464.89 g/mol | C33H68 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW540836250MG
Encompass_Preferred
Tritriacontane is a long-chain linear alkane (C33H68) supplied as a high-purity analytical reagent for research applications. It is used as a chemical standard and reference material in lipid analysis, chromatographic method development, and other analytical workflows.
  • High purity 98.0% suitable for analytical workflows.
  • Well-defined chemical identity: CAS 630-05-7, molecular weight 464.89 g/mol.
  • Available in small, weighed quantities for method development and standards.
  • Stable solid form with long shelf life when stored under recommended conditions.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-83895