Learn More
ATCC Tritrichomonas mobilensis Culberson et al., ATCC 50116 (strain USA:M776)
Supplier:  ATCC 50116
Encompass
ATCC culture of Tritrichomonas mobilensis (strain USA:M776) supplied as an axenic intestinal protozoan culture derived from squirrel monkeys (Saimiri spp.) for laboratory research and characterization.
  • Axenic culture maintained for laboratory research.
  • Cultured in LYI Entamoeba medium.
  • Optimal growth at 35 °C under anaerobic conditions.
  • Strain designation USA:M776, isolated from Saimiri species.
  • Intended for research use in parasitology and protozoan biology.
Catalog No. NC3901749