Learn More
Medchemexpress LLC Trixylyl phosphate | 25155-23-1 | 98.1% | 100 MG

Supplier: Medchemexpress LLC HY122519100MG
Trixylyl phosphate is a triaryl phosphate reagent commonly used as a plasticizer and flame-retardant component in industrial and consumer formulations. It is supplied as a purified research-grade material with supporting documentation for laboratory use.
- High purity: 98.1%.
- Molecular formula C24H27O4P.
- Molecular weight 410.44 g/mol.
- Common applications include plastics and hydraulic fluids.
- Provided with a data sheet, safety data sheet, and certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.