Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma base, 77-86-1, MFCD00004679, 250g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T1503250G
Encompass
Linear Formula NH2C(CH2OH)3, ≥99.9% (titration), Molecular Weight 121.14, bp 219-220 °C/10 mmHg(lit.), mp 167-172 °C (lit.), Primary Standard and Buffer, crystalline, Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Catalog No. 50-165-7245