Learn More
Medchemexpress LLC TrkA-IN-4 | 3026111-74-7 | C27H21F3N4O5 | 1 ML

Supplier: Medchemexpress LLC HY15194910MM/1ML
TrkA-IN-4 is a potent, orally active, and allosteric TrkA inhibitor. It serves as a prodrug of TrkA-IN-3 (IC50=22.4 nM, HY-151948) and exhibits potent antinociceptive effects. This compound is intended for research use only and is not sold to patients.
- Potent, orally active, and allosteric TrkA inhibitor
- Prodrug of TrkA-IN-3
- Exhibits potent antinociceptive effects
- For research use only
- High purity (98.99%)
- Molecular formula: C27H21F3N4O5
- Molecular weight: 538.47
- CAS number: 3026111-74-7
- Recommended storage for powder: -20°C for 3 years
- Recommended storage for in-solvent solution: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.