Learn More
Carnabio Usa Inc TRKB(NTRK2)/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081875
Encompass
TRKB(NTRK2), Cytoplasmic domain, Wild type, Amino Acid 456-822, NP_001018074.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0646