Learn More
ABclonal Technology TrKC Rabbit mAb
SDP
Supplier:  ABclonal Technology A2300220UL
Encompass_Preferred
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family This kinase is a membrane-bound receptor that upon neurotrophin binding phosphorylates itself and members of the MAPK pathway Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position Mutations in this gene have been associated with medulloblastomas secretory breast carcinomas and other cancers Several transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06648