Learn More
Medchemexpress LLC tRNA (cytosine(38)-C(5))-methyltransferase (TRDMT1) | >90.0% | 100 UG

Supplier: Medchemexpress LLC HYP75711100UG
Recombinant human DNMT2 (TRDMT1) expressed in Sf9 insect cells and purified with an N-terminal GST tag. Supplied lyophilized with stabilizing excipients, the protein is intended for research use and requires appropriate storage and reconstitution to preserve activity.
- Expressed in Sf9 insect cells.
- N-terminal GST tag for affinity purification.
- Sequence spans residues M1-E391 (human TRDMT1).
- Lyophilized with Tris, NaCl, GSH, glycerol, trehalose, mannitol, and Tween 80 as protectants.
- Store at -20°C; after reconstitution, stable at 4°C for 1 week or -20°C long term; freeze aliquots for extended storage.
- Purity greater than 90% as determined by reducing SDS-PAGE.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.